missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2,4-dimethyl-1H-pyrrole (>85%), TRC

Product Code. 30137188
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137188 5 g 5g
30133514 25 g 25g
30004027 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137188 Supplier Toronto Research Chemicals Supplier No. TRCE9168805G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-22-6
Molecular Formula C8H13N
Molecular Weight (g/mol) 123.2
Synonym 3-Ethyl-2,4-dimethylpyrrole, Cryptopyrrole, Kryptopyrrol, Kryptopyrrole, NSC 62025, 2,4-Dimethyl-3-ethyl-1H-pyrrole, 2,4-Dimethyl-3-ethylpyrrole
IUPAC Name 3-ethyl-2,4-dimethyl-1H-pyrrole
SMILES CCc1c(C)c[nH]c1C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H13N/c1-4-8-6(2)5-9-7(8)3/h5,9H,4H2,1-3H3
Formula Weight 123.1048
Chemical Name or Material 3-Ethyl-2,4-dimethyl-1H-pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.