missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxymethylene-2,4-pentanedione, 97+%

Product Code. 11441040
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441040 5 g 5g
11451040 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441040 Supplier Thermo Scientific Alfa Aesar Supplier No. B25445.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33884-41-2 | C8H12O3 | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33884-41-2
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
MDL Number MFCD07368652
InChI Key ITTXGKOHFZJUEX-UHFFFAOYSA-N
PubChem CID 240418
IUPAC Name 3-(ethoxymethylidene)pentane-2,4-dione
SMILES CCOC=C(C(C)=O)C(C)=O

Specifications

Boiling Point 140°C to 142°C (15 mmHg)
Refractive Index 1.484
Quantity 5 g
Synonym 3-ethoxymethylene pentane-2,4-dione, 3-ethoxymethylene-2,4-pentanedione, 3-ethoxymethylidene pentane-2,4-dione, acmc-1cmxl, 3-acetyl-4-ethoxy-3-buten-2-one, 3-ethoxymethylene-pentan-2,4-dione, 2,4-pentanedione,3-ethoxymethylene, 2,4-pentanedione, 3-ethoxymethylene, methyl 1-methylcarbonyl-2-ethoxyvinyl ketone
Formula Weight 156.18
Percent Purity ≥98%
Chemical Name or Material 3-Ethoxymethylene-2,4-pentanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.