missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxymethacrolein, 95%, Tech., Thermo Scientific Chemicals

Product Code. 10145030
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42588-57-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00006975
InChI Key KDOAHVPFGIYCEU-AATRIKPKSA-N
Synonym 3-ethoxymethacrolein, 3-ethoxy-2-methylacrylaldehyde, 3-ethoxy-2-methyl-2-propenal, 2e-3-ethoxy-2-methylprop-2-enal, 3-ethoxy-2-methylpropenal, 3-ethoxy-2-methylacroleine, e-2-methyl-3-ethoxypropenal, 2-propenal, 3-ethoxy-2-methyl, e-3-ethoxy-2-methylprop-2-enal
PubChem CID 12278948
IUPAC Name (Z)-3-ethoxy-2-methylprop-2-enal
SMILES CCO\C=C(/C)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.9600g/mL
Boiling Point 78.0°C to 81.0°C (14.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH=C(CH3)CHO
Refractive Index 1.4792 to 1.4812
Quantity 1 g
Beilstein 01, IV, 4082
Specific Gravity 0.96
Formula Weight 114.14
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Ethoxymethacrolein, tech., 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Ethoxymethacrolein
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'