missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethoxycarbonyl)-4-methyl-2-furanpropanoic Acid Ethyl Ester, TRC

Product Code. 30380407
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380407 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380407 Supplier Toronto Research Chemicals Supplier No. TRCE891325250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Ethoxycarbonyl)-4-methyl-2-furanpropanoic Acid Ethyl Ester
CAS 181866-83-1
Molecular Formula C13H18O5
Quantity 250 mg
InChI Formula InChI=1S/C13H18O5/c1-4-16-11(14)7-6-10-12(9(3)8-18-10)13(15)17-5-2/h8H,4-7H2,1-3H3
SMILES O=C(OCC)C1=C(CCC(OCC)=O)OC=C1C
IUPAC Name ethyl 2-(3-ethoxy-3-oxopropyl)-4-methylfuran-3-carboxylate
Molecular Weight (g/mol) 254.28
Formula Weight 254.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.