missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzonitrile, 95%

Product Code. 11491618
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491618 25 g 25g
11481618 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491618 Supplier Thermo Scientific Alfa Aesar Supplier No. A13908.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25117-75-3 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25117-75-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00001805
InChI Key PFNAUBYHTWDKBO-UHFFFAOYSA-N
Synonym benzonitrile, 3-ethoxy, benzonitrile, m-ethoxy, m-ethoxy benzonitrile, acmc-20an4n, 3-ethoxybenzenecarbonitrile, 3-ethoxybenzoic acid nitrile
PubChem CID 91283
IUPAC Name 3-ethoxybenzonitrile
SMILES CCOC1=CC=CC(=C1)C#N

Specifications

Density 1.047
Boiling Point 241°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 25 g
UN Number UN3276
Formula Weight 147.18
Percent Purity 95%
Chemical Name or Material 3-Ethoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.