missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxyacrylic acid, 95%, Thermo Scientific™

Product Code. 10306954
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306954 5 g Amber Glass Bottle 5g
10289822 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306954 Supplier Thermo Scientific Maybridge Supplier No. RH00924DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6192-01-4
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
InChI Key SYMAGJYJMLUEQE-ONEGZZNKSA-N
Synonym 3-ethoxyacrylic acid, e-3-ethoxyacrylic acid, 3-ethoxyprop-2-enoic acid, 3-ethyoxyacrylic acid, 3-ethoxy-2-propenoic acid, 2e-3-ethoxyprop-2-enoic acid, 2e-3-ethoxyacrylic acid, 3-ethoxyacrylicacid, pubchem20958, e-3-ethoxy-acrylic acid
PubChem CID 5709609

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 116.12
Percent Purity 95%
Chemical Name or Material 3-Ethoxyacrylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.