missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15440276
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440276 1 g 1g
15450276 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440276 Supplier Thermo Scientific Alfa Aesar Supplier No. H52989.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 871332-96-6 | Molecular Formula: C9H10BF3O3 | Molecular Weight: 233.98 g/mol

Reactant involved in suzuki-miyaura cross-coupling reactions, aerobic oxidative cross-coupling, rhodium-catalyzed addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant involved in suzuki-miyaura cross-coupling reactions, aerobic oxidative cross-coupling, rhodium-catalyzed addition reactions.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871332-96-6
Molecular Formula C9H10BF3O3
Molecular Weight (g/mol) 233.98
MDL Number MFCD08056361
InChI Key ATZRXKHRSOTBAM-UHFFFAOYSA-N
Synonym 3-ethoxy-5-trifluoromethyl phenylboronic acid, 3-ethoxy-5-trifluoromethyl benzeneboronic acid, boronic acid,b-3-ethoxy-5-trifluoromethyl phenyl, 3-ethoxy-5-trifluoromethylphenylboronic acid, 3-ethoxy-5-trifluoromethyl phenyl boronic acid, acmc-209qhp, 3-ethoxy-5-trifluoromethyl benzeneboronicacid
PubChem CID 44717589
IUPAC Name [3-ethoxy-5-(trifluoromethyl)phenyl]boronic acid
SMILES CCOC1=CC(=CC(=C1)C(F)(F)F)B(O)O

Specifications

Melting Point 173°C to 176°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 233.98
Percent Purity 98%
Chemical Name or Material 3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.