missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-5-methylphenylboronic acid, TRC

Product Code. 30384924
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384924 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384924 Supplier Toronto Research Chemicals Supplier No. TRCE894888100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Ethoxy-5-methylphenylboronic acid
CAS 1256346-05-0
Molecular Formula C9H13BO3
Quantity 100 mg
InChI Formula InChI=1S/C9H13BO3/c1-3-13-9-5-7(2)4-8(6-9)10(11)12/h4-6,11-12H,3H2,1-2H3
SMILES CCOc1cc(C)cc(c1)B(O)O
Molecular Weight (g/mol) 180.01
Formula Weight 180.0958
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.