missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-methoxybenzaldehyde, 99%

Product Code. 11496476
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496476 5 g 5g
11406486 25 g 25g
11416486 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496476 Supplier Thermo Scientific Alfa Aesar Supplier No. B24762.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1131-52-8 | C10H12O3 | 180.203 g/mol

3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Store in a tightly sealed container. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-52-8
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00010128
InChI Key VAMZHXWLGRQSJS-UHFFFAOYSA-N
Synonym 3-ethoxy-4-methoxy-benzaldehyde, benzaldehyde, 3-ethoxy-4-methoxy, pubchem13135, 3-ethoxy-p-anisaldehyde, acmc-1ca82, 3-ethoxy-4methoxybenzaldehyde, 3ethoxy-4-methoxybenzaldehyde, ksc183c6j, 3-ethoxy-4-methoxybenzaldehyd, 4-methoxy-3-ethoxybenzaldehyde
PubChem CID 244728
IUPAC Name 3-ethoxy-4-methoxybenzaldehyde
SMILES CCOC1=C(C=CC(=C1)C=O)OC

Specifications

Melting Point 51°C to 53°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material 3-Ethoxy-4-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.