missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-hydroxybenzaldehyde, 98%

Product Code. 11389489
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389489 25 g 25g
11404057 100 g 100g
11414057 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389489 Supplier Thermo Scientific Alfa Aesar Supplier No. A19478.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-32-4 | C9H10O3 | 166.18 g/mol

It plays an important role as an antioxidant and is also used as a flavoring agent, in perfumes, and in organic synthesis. It holds anti-angiogenic, anti-inflammatory and antinociceptive properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role as an antioxidant and is also used as a flavoring agent, in perfumes, and in organic synthesis. It holds anti-angiogenic, anti-inflammatory and antinociceptive properties.

Solubility
Slightly soluble in water. Soluble in ethanol, ether, benzene.

Notes
Air & light sensitive. Protect from light. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-32-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00006944
InChI Key CBOQJANXLMLOSS-UHFFFAOYSA-N
Synonym ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal
PubChem CID 8467
ChEBI CHEBI:48408
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
SMILES CCOC1=CC(C=O)=CC=C1O

Specifications

Melting Point 75°C to 79°C
Boiling Point 285°C
Flash Point 127°C (260°F)
Quantity 25 g
Beilstein 1073761
Sensitivity Air and light sensitive
Merck Index 14,3859
Solubility Information Slightly soluble in water. Soluble in ethanol,ether,benzene.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3-Ethoxy-4-hydroxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethoxy-4-hydroxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: 3-Ethoxy-4-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.