missing translation for 'onlineSavingsMsg'
Learn More

3-epi-25-Hydroxy Vitamin D3, TRC

Product Code. 30398644
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398644 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30398644 Supplier Toronto Research Chemicals Supplier No. TRCH9958601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-epi-25-Hydroxyvitamin D3
CAS 73809-05-9
Purity Grade Notes HPLC
Molecular Formula C27 H44 O2
Quantity 1 mg
Synonym 1H-Indene-1-pentanol, octahydro-4-[(2Z)-2-[(5R)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-α,α,ε,7a-tetramethyl-, (εR,1R,3aS,4E,7aR)-, 9,10-Secocholesta-5,7,10(19)-triene-3,25-diol, (3α,5Z,7E)- (9CI), (εR,1R,3aS,4E,7aR)-Octahydro-4-[(2Z)-2-[(5R)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-α,α,ε,7a-tetramethyl-1H-indene-1-pentanol, 3-epi-Calcifediol, 3-epi-25-Hydroxycholecalciferol, 3-epi-25-Hydroxyitamin D3, 3-epi-25-Hydroxyvitamin D3, Vitamin D3-3-epi-25-hydroxy
InChI Formula InChI=1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23-,24-,25+,27-/m1/s1
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@H](O)CCC3=C)\CCC[C@]12C
IUPAC Name (1R,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molecular Weight (g/mol) 400.64
Formula Weight 400.3341
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.