missing translation for 'onlineSavingsMsg'
Learn More

3-(Diphenylphosphino)benzenesulfonic Acid Sodium Salt, TRC

Product Code. 30372682
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372682 10 mg
30371532 100 mg
30384962 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372682 Supplier Toronto Research Chemicals Supplier No. TRCD49492010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63995-75-5
Molecular Formula C18H14NaO3PS
Molecular Weight (g/mol) 364.33
Synonym Diphenyl(3-sulfophenyl)phosphine Sodium Salt, Diphenylphosphinobenzene-m-sulfonic Acid Sodium Salt, Monosodium Mono(m-sulfonatophenyl)diphenylphosphine, Sodium (Diphenylphosphino)benzene-3-sulfonate, Sodium diphenyl(3-sulfonatophenyl)phosphine Dihydrate, TPPMS, [3-(Sodium Sulfonato)phenyl]diphenylphosphine, 3-Sulfonatophenyl)diphenylphosphane Sodium Salt, (m-Sulfonatophenyl)diphenylphosphine Sodium Salt, (m-Sulfophenyl)diphenylphosphine Sodium Salt
SMILES [Na+].[O-]S(=O)(=O)c1cccc(c1)P(c2ccccc2)c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H15O3PS.Na/c19-23(20,21)18-13-7-12-17(14-18)22(15-8-3-1-4-9-15)16-10-5-2-6-11-16;/h1-14H,(H,19,20,21);/q;+1/p-1
Chemical Name or Material 3-(Diphenylphosphino)benzenesulfonic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.