missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylsulfamoyl)benzeneboronic acid, 98%

Product Code. 15409617
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409617 250 mg 250mg
15499607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409617 Supplier Thermo Scientific Alfa Aesar Supplier No. H52838.MD
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 871329-59-8 | C8H12BNO4S | 229.057 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871329-59-8
Molecular Formula C8H12BNO4S
Molecular Weight (g/mol) 229.057
MDL Number MFCD07363747
InChI Key SGERXKCDWJPIOS-UHFFFAOYSA-N
Synonym 3-n,n-dimethylsulfamoyl phenyl boronic acid, n,n-dimethyl 3-boronobenzenesulfonamide, 3-dimethylsulfamoyl phenyl boronic acid, 3-n,n-dimethylsulphonamido benzeneboronic acid, 3-dimethylsulfamoyl phenylboronic acid, 3-dimethylsulfamoyl benzeneboronic acid, 3-n,n-dimethylsulfonamidophenyl boronic acid, boronic acid, 3-dimethylamino sulfonyl phenyl, boronic acid,b-3-dimethylamino sulfonyl phenyl, acmc-209qg4
PubChem CID 44119634
IUPAC Name [3-(dimethylsulfamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)N(C)C)(O)O

Specifications

Melting Point 88°C to 92°C
Quantity 250 mg
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material 3-(Dimethylsulfamoyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.