missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamlno)phenol, TRC

Product Code. 30374910
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374910 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374910 Supplier Toronto Research Chemicals Supplier No. TRCD4613005G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 3-(Dimethylamino)phenol
CAS 99-07-0
Molecular Formula C8H11NO
Quantity 5 g
Synonym Phenol, 3-(dimethylamino)-, Phenol, m-(dimethylamino)- (6CI,7CI,8CI), 3-(Dimethylamino)phenol, (3-Hydroxyphenyl)dimethylamine, 3-Hydroxy-N,N-dimethylaniline, 3-N,N-Dimethylaminophenol, N,N-Dimethyl-3-aminophenol, N,N-Dimethyl-m-aminophenol, NSC 62017, m-(Dimethylamino)phenol, m-(N,N-Dimethylamino)phenol
InChI Formula InChI=1S/C8H11NO/c1-9(2)7-4-3-5-8(10)6-7/h3-6,10H,1-2H3
SMILES CN(C)c1cccc(O)c1
IUPAC Name 3-(dimethylamino)phenol
Molecular Weight (g/mol) 137.18
Formula Weight 137.0841
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.