missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)propionic acid hydrochloride, TRC

Product Code. 30381177
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381177 250 mg -
30384523 500 mg -
30370412 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381177 Supplier Toronto Research Chemicals Supplier No. TRCD456735250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14788-12-6
Molecular Formula C5H11NO2 . HCl
Molecular Weight (g/mol) 153.61
Synonym 3-(Dimethylamino)propionic Acid Hydrochloride, 3-(N,N-Dimethylamino)propionic Acid Hydrochloride, 3-Dimethylaminopropanoic Acid HydrochlorideN,N-Dimethyl-beta-alanine Hydrochloride, beta-(Dimethylamino)propionic Acid Hydrochloride
IUPAC Name 3-(dimethylamino)propanoic acid;hydrochloride
SMILES Cl.CN(C)CCC(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H11NO2.ClH/c1-6(2)4-3-5(7)8;/h3-4H2,1-2H3,(H,7,8);1H
Formula Weight 153.0557
Chemical Name or Material 3-(Dimethylamino)propionic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.