missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-2-methylpropyl chloride hydrochloride, 98%, Thermo Scientific™

Product Code. 11362366
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362366 500 g 500g
11342366 25 g 25g
11352366 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362366 Supplier Thermo Scientific Alfa Aesar Supplier No. L06091.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4261-67-0
Molecular Formula C6H15Cl2N
Molecular Weight (g/mol) 172.093
MDL Number MFCD00012514
InChI Key SOMIBONUMGNAEP-UHFFFAOYSA-N
Synonym 3-dimethylamino-2-methylpropyl chloride hydrochloride, 3-chloro-n,n,2-trimethylpropan-1-amine hydrochloride, 3-chloro-2-methylpropyl dimethyl ammonium chloride, 3-chloro-2-methylpropyl dimethylamine hydrochloride, acmc-20an51, 3-dimethylamino-2-methylpropyl chloride hcl, 3-chloro-n,n,2-trimethylpropylamine hydrochloride, 2-methyl-3-dimethylaminopropylchloride hydrochloride, 3-dimethylamino-2-methylpropylchloride hydrochloride, 1-dimethylamino-2-methyl-3-chloropropane hydrochloride
PubChem CID 3083841
IUPAC Name 3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
SMILES CC(CN(C)C)CCl.Cl

Specifications

Melting Point 167°C to 170°C
Quantity 500 g
Beilstein 4444785
Sensitivity Hygroscopic
Formula Weight 172.1
Percent Purity 98%
Chemical Name or Material 3-Dimethylamino-2-methylpropyl chloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.