missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-1-propanol, 99%

Product Code. 10529420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10529420 500 g Glass bottle 500g
10496910 5 g Glass bottle 5g
10406920 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10529420 Supplier Thermo Scientific Acros Supplier No. 115885000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3179-63-3 | C5H13NO | 103.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3179-63-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00002945
InChI Key PYSGFFTXMUWEOT-UHFFFAOYSA-N
Synonym 3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol
PubChem CID 76646
IUPAC Name 3-(dimethylamino)propan-1-ol
SMILES CN(C)CCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless to Yellow
Density 0.8860g/mL
Boiling Point 159.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2N(CH2)3OH
Refractive Index 1.4350 to 1.4370
Quantity 500 g
Beilstein 04, I, 433
Specific Gravity 0.886
Solubility Information Solubility in water: soluble
Viscosity 6.85 mPa.s (20°C)
Formula Weight 103.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Dimethylamino-1-propanol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Dimethylamino-1-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.