missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-1-propanol, 99%, Thermo Scientific Chemicals

Product Code. 11425163
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425163 100 mL 100mL
11435163 500 mL 500mL
11445163 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425163 Supplier Thermo Scientific Alfa Aesar Supplier No. A12800.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylamino-1-propanol, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3179-63-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00002945
InChI Key PYSGFFTXMUWEOT-UHFFFAOYSA-N
Synonym 3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol
PubChem CID 76646
IUPAC Name 3-(dimethylamino)propan-1-ol
SMILES CN(C)CCCO

Specifications

Melting Point -35°C
Density 0.88
Boiling Point 162°C to 164°C
Flash Point 54°C (129°F)
Linear Formula (CH3)2N(CH2)3OH
Refractive Index 1.436
Quantity 100 mL
UN Number UN2734
Beilstein 1732032
Sensitivity Air Sensitive
Solubility Information It is miscible with water.
Formula Weight 103.16
Percent Purity 99%
Chemical Name or Material 3-Dimethylamino-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.