missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethoxy)benzenesulfonyl Chloride, TRC

Product Code. 30376681
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376681 250 mg
30384614 100 mg
30383972 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376681 Supplier Toronto Research Chemicals Supplier No. TRCD447713250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-38-8
Molecular Formula C7H5ClF2O3S
Molecular Weight (g/mol) 242.63
SMILES FC(F)Oc1cccc(c1)S(=O)(=O)Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H5ClF2O3S/c8-14(11,12)6-3-1-2-5(4-6)13-7(9)10/h1-4,7H
Formula Weight 241.9616
Chemical Name or Material 3-(Difluoromethoxy)benzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.