missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylamino-1-propyne, 98+%

Product Code. 11388685
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388685 5 g 5g
11398685 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388685 Supplier Thermo Scientific Alfa Aesar Supplier No. L12237.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4079-68-9 | C7H13N | 111.188 g/mol

3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

Solubility
Soluble in acid. Insoluble in water.

Notes
Incompatible with heat, light, carbondioxide, heavy metal powders. Air & Light Sensitive. Store in the dark. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4079-68-9
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.188
MDL Number MFCD00039881
InChI Key JZJXKEWVUBVOEH-UHFFFAOYSA-N
Synonym 1-diethylamino-2-propyne, diethylpropargylamine, 2-propyn-1-amine, n,n-diethyl, n,n-diethylpropargylamine, n,n-diethyl-2-propynylamine, 3-diethylamino propyne, 3-diethylamino-1-propyne, 3-n,n-diethylamino propyne, 1-n,n-diethylamino-2-propyne, 2-propynylamine, n,n-diethyl
PubChem CID 20010
IUPAC Name N,N-diethylprop-2-yn-1-amine
SMILES CCN(CC)CC#C

Specifications

Density 0.804
Boiling Point 119°C to 120°C
Flash Point 20°C (68°F)
Odor Amine-like
Linear Formula (CH3CH2)2NCH2C≡CH
Refractive Index 1.4305
Quantity 5 g
UN Number UN2733
Beilstein 1098468
Sensitivity Air and light sensitive
Solubility Information Soluble in acid. Insoluble in water.
Formula Weight 111.19
Percent Purity ≥98%
Chemical Name or Material 3-Diethylamino-1-propyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.