missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylamino-1-propanol, 95%

Product Code. 10414272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10414272 5 g 5g
10651785 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414272 Supplier Thermo Scientific Acros Supplier No. 165410050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 622-93-5 | C7H17NO | 131.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-93-5
Molecular Formula C7H17NO
Molecular Weight (g/mol) 131.22
MDL Number MFCD00002947
InChI Key WKCYFSZDBICRKL-UHFFFAOYSA-N
Synonym 3-diethylamino-1-propanol, 3-diethylamino propan-1-ol, 1-propanol, 3-diethylamino, 3-diethylamino propanol, 3-diethylamino propyl alcohol, n,n-diethyl-3-hydroxypropylamine, diethylpropanolamine, unii-u57yow5553, gamma-diaethylaminopropanol german
PubChem CID 12159
IUPAC Name 3-(diethylamino)propan-1-ol
SMILES CCN(CC)CCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 81°C to 83°C (15.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2N(CH2)3OH
Refractive Index 1.4425 to 1.4445
Quantity 5 g
Beilstein 04,288
Specific Gravity 0.88
Formula Weight 131.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Diethylamino-1-propanol, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.