missing translation for 'onlineSavingsMsg'
Learn More

3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-diethylpropan-1-amine Hydrochloride (E/Z Mixture), TRC

Product Code. 30380320
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380320 500 mg 500mg
30386800 250 mg 250mg
30378739 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380320 Supplier Toronto Research Chemicals Supplier No. TRCD418705500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-diethylpropan-1-amine Hydrochloride (E/Z Mixture)
Analyte or Component Names 3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-diethylpropan-1-amine Hydrochloride (E/Z Mixture)
Shipping Condition Room Temperature
Molecular Formula C21H25NO • HCl
InChI Formula InChI=1S/C21H25NO.ClH/c1-3-22(4-2)15-9-13-19-18-11-6-5-10-17(18)16-23-21-14-8-7-12-20(19)21;/h5-8,10-14H,3-4,9,15-16H2,1-2H3;1H/b19-13-;
Solution Type Neat
SMILES CCN(CC)CC/C=C1C2=CC=CC=C2OCC3=C\1C=CC=C3.Cl
IUPAC Name 3-(dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-diethylpropan-1-amine hydrogen chloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 343.89
Quantity 500 mg
Formula Weight 343.17
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.