missing translation for 'onlineSavingsMsg'
Learn More

3-Descyano Febuxostat, TRC

Product Code. 30357908
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357908 100 mg 100mg
30362348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357908 Supplier Toronto Research Chemicals Supplier No. TRCD289705100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1206550-99-3
Molecular Formula C15H17NO3S
Molecular Weight (g/mol) 291.37
Synonym 2-(4-Isobutoxyphenyl)-4-methylthiazole-5-carboxylic Acid
SMILES CC(C)COC1=CC=C(C2=NC(C)=C(C(O)=O)S2)C=C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H17NO3S/c1-9(2)8-19-12-6-4-11(5-7-12)14-16-10(3)13(20-14)15(17)18/h4-7,9H,8H2,1-3H3,(H,17,18)
Percent Purity >95%
Chemical Name or Material 3-Descyano Febuxostat
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.