missing translation for 'onlineSavingsMsg'
Learn More

3-Deschloro-4-desfluoro-4-chloro-3-fluoro Gefitinib, TRC

Product Code. 30005836
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005836 100 mg 100mg
17922535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005836 Supplier Toronto Research Chemicals Supplier No. TRCD289540100MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603814-04-5
Molecular Formula C22 H24 Cl F N4 O3
Molecular Weight (g/mol) 446.9
Synonym 4-Quinazolinamine, N-(4-chloro-3-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-, Gefitinib Imp. B (EP), N-(4-Chloro-3-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-4-quinazolinamine, N-(4-Chloro-3-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]quinazolin-4-amine, 3-Deschloro-4-desfluoro-4-chloro-3-fluoro Gefitinib
IUPAC Name N-(4-chloro-3-fluorophenyl)-7-methoxy-6-(3-morpholin-4-ylpropoxy)quinazolin-4-amine
SMILES COc1cc2ncnc(Nc3ccc(Cl)c(F)c3)c2cc1OCCCN4CCOCC4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H24ClFN4O3/c1-29-20-13-19-16(12-21(20)31-8-2-5-28-6-9-30-10-7-28)22(26-14-25-19)27-15-3-4-17(23)18(24)11-15/h3-4,11-14H,2,5-10H2,1H3,(H,25,26,27)
Formula Weight 446.1521
Percent Purity >95%
Chemical Name or Material 3-Deschloro-4-desfluoro-4-chloro-3-fluoro Gefitinib
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.