missing translation for 'onlineSavingsMsg'
Learn More

3-Deschloro-4-chloro Bupropion Hydrochloride, TRC

Product Code. 30366218
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366218 50 mg 50mg
30359909 10 mg 10mg
30353418 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366218 Supplier Toronto Research Chemicals Supplier No. TRCD28192550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1049718-72-0
Molecular Formula C13 H18 Cl N O . Cl H
Molecular Weight (g/mol) 276.2
Synonym 2-(tert-Butylamino)-4'-chloropropiophenone Hydrochloride
IUPAC Name 2-(tert-butylamino)-1-(4-chlorophenyl)propan-1-one;hydrochloride
SMILES Cl.CC(NC(C)(C)C)C(=O)c1ccc(Cl)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H18ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-5-7-11(14)8-6-10;/h5-9,15H,1-4H3;1H
Formula Weight 275.0844
Chemical Name or Material 2-(tert-Butylamino)-4'-chloropropiophenone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.