missing translation for 'onlineSavingsMsg'
Learn More

3-Deschloro-3,4-difluoro Gefitinib, TRC

Product Code. 30012287
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012287 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012287 Supplier Toronto Research Chemicals Supplier No. TRCD293125500MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 184475-50-1
Molecular Formula C22 H24 F2 N4 O3
Quantity 500 mg
Synonym 3-Deschloro-3,4-difluoro Gefitinib;
InChI Formula InChI=1S/C22H24F2N4O3/c1-29-20-13-19-16(12-21(20)31-8-2-5-28-6-9-30-10-7-28)22(26-14-25-19)27-15-3-4-17(23)18(24)11-15/h3-4,11-14H,2,5-10H2,1H3,(H,25,26,27)
SMILES COc1cc2ncnc(Nc3ccc(F)c(F)c3)c2cc1OCCCN4CCOCC4
IUPAC Name N-(3,4-difluorophenyl)-7-methoxy-6-(3-morpholin-4-ylpropoxy)quinazolin-4-amine
Molecular Weight (g/mol) 430.448
Formula Weight 430.182
Chemical Name or Material 3-Deschloro-3,4-difluoro Gefitinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.