missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Deoxyuridine

Product Code. 15446959
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446959 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446959 Supplier Thermo Scientific Chemicals Supplier No. J65531.MB
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7057-27-4 | C9H12N2O5 | 228.204 g/mol

A potential anti-cancer and anti-viral agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7057-27-4
Color White
Molecular Formula C9H12N2O5
MDL Number MFCD00079153
Quantity 25 mg
Synonym 3'-deoxyuridine, uridine, 3'-deoxy, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 3-deoxyuridine, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, uridine, 3/'-deoxy, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl oxolan-2-yl pyrimidine-2,4-dione
Solubility Information Soluble in DMSO,methanol,or water; Also soluble in dichloromethane /n
InChI Key QOXJRLADYHZRGC-SHYZEUOFSA-N
SMILES C1C(OC(C1O)N2C=CC(=O)NC2=O)CO
IUPAC Name 1-[(2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 228.204
PubChem CID 145984
Formula Weight 228.21
Physical Form Powder
Chemical Name or Material 3'-Deoxyuridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.