missing translation for 'onlineSavingsMsg'
Learn More

3-Deoxyaconitine, TRC

Product Code. 30357423
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357423 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357423 Supplier Toronto Research Chemicals Supplier No. TRCD23155310MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Deoxyaconitine
CAS 3175-95-9
Molecular Formula C34 H47 N O10
Quantity 10 mg
Synonym Deoxyaconitine, (1α,6α,14α,15α,16β)-20-Ethyl-1,6,16-trimethoxy-4-(methoxymethyl)aconitane-8,13,14,15-tetrol 8-Acetate 14-Benzoate
InChI Formula InChI=1S/C34H47NO10/c1-7-35-16-31(17-40-3)14-13-21(41-4)33-20-15-32(39)28(44-30(38)19-11-9-8-10-12-19)22(20)34(45-18(2)36,27(37)29(32)43-6)23(26(33)35)24(42-5)25(31)33/h8-12,20-29,37,39H,7,13-17H2,1-6H3/t20-,21+,22-,23+,24+,25-,26+,27+,28-,29+,31+,32-,33+,34-/m1/s1
SMILES O(C)[C@@H]1[C@]23[C@]4([C@]([C@H](OC)[C@@]2([C@](COC)(CN4CC)CC1)[H])([C@]5(OC(C)=O)[C@@]6([C@]3(C[C@@](O)([C@@H]6OC(=O)C7=CC=CC=C7)[C@@H](OC)[C@@H]5O)[H])[H])[H])[H]
IUPAC Name [(1S,2R,3R,4R,5R,6S,7S,8R,9R,10S,13S,16S,17R,18R)-8-acetoxy-11-ethyl-5,7-dihydroxy-6,16,18-trimethoxy-13-(methoxymethyl)-11-azahexacyclo[7.7.2.12,5.01,10.03,8.013,17]nonadecan-4-yl] benzoate
Molecular Weight (g/mol) 629.73768
Formula Weight 629.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.