missing translation for 'onlineSavingsMsg'
Learn More

3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals

Product Code. 15497489
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497489 5 mg 5mg
15407499 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497489 Supplier Thermo Scientific Alfa Aesar Supplier No. J66223.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents. Also used for cross-linking studies as it readily reacts with protein amino groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents
  • Also used for cross-linking studies as it readily reacts with protein amino groups
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4084-27-9
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD01723101
InChI Key ZGCHLOWZNKRZSN-NTSWFWBYSA-N
Synonym 3-deoxyglucosone, 3-deoxy-d-glucosone, 3-deoxy-d-erythro-hexos-2-ulose, d-3-deoxyglucosone, 3-deoxy-d-erythro-hexosulose, 3-deoxy-d-fructose, ccris 4272, 3-deoxy-2-ketoglucose, d-erythro-hexos-2-ulose, 3-deoxy, unii-exv5374vey
PubChem CID 114839
ChEBI CHEBI:60777
IUPAC Name (4S,5R)-4,5,6-trihydroxy-2-oxohexanal
SMILES C(C(C(CO)O)O)C(=O)C=O

Specifications

Color Yellow
Quantity 5 mg
Solubility Information Soluble in water,methanol or hot ethanol.
Formula Weight 162.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Deoxy-D-glucosone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.