missing translation for 'onlineSavingsMsg'
Learn More

3’-Deoxy-3’-fluoro Thymidine, TRC

Product Code. 30361821
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361821 50 mg 50mg
30359239 250 mg 250mg
30357911 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361821 Supplier Toronto Research Chemicals Supplier No. TRCD23547550MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25526-93-6
Molecular Formula C10H13FN2O4
Molecular Weight (g/mol) 244.22
SMILES CC1=CN([C@H]2C[C@H](F)[C@@H](CO)O2)C(=O)NC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13FN2O4/c1-5-3-13(10(16)12-9(5)15)8-2-6(11)7(4-14)17-8/h3,6-8,14H,2,4H2,1H3,(H,12,15,16)/t6-,7+,8+/m0/s1
Chemical Name or Material 3'-Deoxy-3'-fluoro Thymidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.