missing translation for 'onlineSavingsMsg'
Learn More

3'-Deoxy-2',3'-didehydrothymidine, 98%

Product Code. 15246058 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15246058 5 g 5g
15288155 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15246058 Supplier Thermo Scientific Acros Supplier No. 454940050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3056-17-5 | C10H12N2O4 | 224.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3056-17-5
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.22
MDL Number MFCD00132921
InChI Key XNKLLVCARDGLGL-HGXVMFPFNA-N
Synonym stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine
PubChem CID 18283
ChEBI CHEBI:63581
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O

Specifications

Melting Point 162.0°C to 169.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Merck Index 158929
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 3'-Deoxy-2',3'-didehydrothymidine
compliance-icons compliance-icons
Product Identifier
  • 3'-Deoxy-2',3'-didehydrothymidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.