missing translation for 'onlineSavingsMsg'
Learn More

3-Dehydroxy Chlorthalidone, TRC

Product Code. 30135142
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135142 100 mg 100mg
30005794 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135142 Supplier Toronto Research Chemicals Supplier No. TRCD230540100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82875-49-8
Molecular Formula C14 H11 Cl N2 O3 S
Molecular Weight (g/mol) 322.77
Synonym Benzenesulfonamide, 2-chloro-5-(2,3-dihydro-3-oxo-1H-isoindol-1-yl)-, 2-Chloro-5-(2,3-dihydro-3-oxo-1H-isoindol-1-yl)benzenesulfonamide, 2-Chloro-5-[(1RS)-3-oxo-2,3-dihydro-1H-isoindol-1-yl]benzenesulfonamide, 3-Dehydroxy Chlorthalidone, Chlortalidone Imp. E (EP)
IUPAC Name 2-chloro-5-(3-oxo-1,2-dihydroisoindol-1-yl)benzenesulfonamide
SMILES NS(=O)(=O)c1cc(ccc1Cl)C2NC(=O)c3ccccc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H11ClN2O3S/c15-11-6-5-8(7-12(11)21(16,19)20)13-9-3-1-2-4-10(9)14(18)17-13/h1-7,13H,(H,17,18)(H2,16,19,20)
Formula Weight 322.0179
Chemical Name or Material 3-Dehydroxy Chlorthalidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.