missing translation for 'onlineSavingsMsg'
Learn More

3-Decyn-1-ol, 97%

Product Code. 11445776
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11445776

Brand: Thermo Scientific Alfa Aesar B20837.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51721-39-2 | C10H18O | 154.253 g/mol

3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3?-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3′-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

Solubility
Sparingly soluble in water (0.21 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51721-39-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00040917
InChI Key YGEQBZUDPQQIFI-UHFFFAOYSA-N
Synonym 3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
PubChem CID 103940
IUPAC Name dec-3-yn-1-ol
SMILES CCCCCCC#CCCO

Specifications

Density 0.871
Boiling Point 129°C to 130°C (20 mmHg)
Flash Point 106°C (222°F)
Refractive Index 1.459
Quantity 25 g
Beilstein 1750909
Solubility Information Sparingly soluble in water (0.21g/L) (25°C).
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material 3-Decyn-1-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.