missing translation for 'onlineSavingsMsg'
Learn More

3-Deazaneplanocin A hydrochloride, MedChemExpress

Product Code. 16447737
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16447737

Brand: Medchem Express HY12186

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Deazaneplanocin A hydrochloride (DZNep hydrochloride) is a potent histone methyltransferase EZH2 inhibitor. 3-Deazaneplanocin A hydrochlorideis a potent S-adenosylhomocysteine hydrolase (AHCY) inhibitor. 3-Deazaneplanocin A hydrochloride also has anti-orthopoxvirus and anti-variola activities.

3-Deazaneplanocin A hydrochloride (DZNep hydrochloride) is a potent histone methyltransferase EZH2 inhibitor. 3-Deazaneplanocin A hydrochlorideis a potent S-adenosylhomocysteine hydrolase (AHCY) inhibitor. 3-Deazaneplanocin A hydrochloride also has anti-orthopoxvirus and anti-variola activities.

Specifications

Chemical Name or Material 3-Deazaneplanocin A hydrochloride
CAS 120964-45-6
Color Light Brown
Purity Grade Notes Research
Molecular Formula C12H15ClN4O3
Quantity 5 mg
Synonym DZNep hydrochloride NSC 617989 hydrochloride 3-Deazaneplanocin hydrochloride
Solubility Information H2O : 50 mg/mL (167.38 mM; Need ultrasonic)
SMILES O[C@@H]1[C@H](O)C(CO)=C[C@H]1N2C=NC3=C2C=CN=C3N.[H]Cl
Molecular Weight (g/mol) 298.73
Formula Weight 298.73
Percent Purity 98.37%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.