missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyclopropylsulfamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15479177
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479177 1 g 1g
15419805 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479177 Supplier Thermo Scientific Alfa Aesar Supplier No. H52441.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-28-6
Molecular Formula C9H12BNO4S
Molecular Weight (g/mol) 241.07
MDL Number MFCD08436035
InChI Key WIZSPBQIRRAOMO-UHFFFAOYSA-N
Synonym 3-n-cyclopropylsulfamoyl phenyl boronic acid, n-cyclopropyl 3-boronobenzenesulfonamide, boronicacid, b-3-cyclopropylamino sulfonyl phenyl, 3-cyclopropylsulfamoyl phenylboronic acid, 3-cyclopropylsulfamoyl benzeneboronic acid, 3-n-cyclopropylsulfamoyl phenylboronic acid, n-cyclopropyl-3-boronobenzenesulfonamide, acmc-209r8z, 3-cyclopropylsulfamoyl phenyl boronic acid, 3-n-cyclopropylsulfamoyl phenyl boronicacid
PubChem CID 44119335
IUPAC Name [3-(cyclopropylsulfamoyl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC=C1)S(=O)(=O)NC1CC1

Specifications

Melting Point 126°C to 128°C
Quantity 1 g
Formula Weight 241.07
Percent Purity 98%
Chemical Name or Material 3-(Cyclopropylsulfamoyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.