missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropylmethoxy-4-difluoromethoxybenzoic Acid, TRC

Product Code. 30366126
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30366126 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366126 Supplier Toronto Research Chemicals Supplier No. TRCC9887201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzoic Acid
CAS 162401-62-9
Molecular Formula C12 H12 F2 O4
Quantity 1 g
Synonym 3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzoic Acid, 3-Cyclopropylmethoxy-4-difluoromethoxybenzoic Acid
InChI Formula InChI=1S/C12H12F2O4/c13-12(14)18-9-4-3-8(11(15)16)5-10(9)17-6-7-1-2-7/h3-5,7,12H,1-2,6H2,(H,15,16)
SMILES OC(=O)c1ccc(OC(F)F)c(OCC2CC2)c1
IUPAC Name 3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
Molecular Weight (g/mol) 258.22
Formula Weight 258.0704
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.