missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropyl-3-oxopropanoic Acid Methyl Ester, TRC

Product Code. 30361095
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361095 Supplier Toronto Research Chemicals Supplier No. TRCC9887751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Cyclopropyl-3-oxopropanoic Acid Methyl Ester
CAS 32249-35-7
Molecular Formula C7H10O3
Quantity 1 g
Synonym Methyl 3-Cyclopropyl-3-oxopropionate, Methyl 2-(Cyclopropylcarbonyl)acetate, beta-Oxo-cyclopropanepropanoic Acid Methyl Ester
InChI Formula InChI=1S/C7H10O3/c1-10-7(9)4-6(8)5-2-3-5/h5H,2-4H2,1H3
SMILES COC(=O)CC(=O)C1CC1
IUPAC Name methyl 3-cyclopropyl-3-oxopropanoate
Molecular Weight (g/mol) 142.15
Formula Weight 142.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.