missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Product Code. 15491099
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100114-57-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD04970938,MFCD06739298
InChI Key TXWDVWSJMDFNQY-UHFFFAOYSA-N
PubChem CID 7017816
IUPAC Name 5-cyclopropyl-1H-pyrazole
SMILES C1CC1C1=CC=NN1

Specifications

Color Colorless
Quantity 1 g
Beilstein 1561659
Synonym 3-cyclopropyl-1h-pyrazole, 5-cyclopropylpyrazole, 1h-pyrazole, 3-cyclopropyl, 3-cyclopropylpyrazole, 1h-pyrazole,3-cyclopropyl, 3-cyclopropyl-2h-pyrazole, 3-cyclopropyl-1h-pyrazole 97+%, acmc-2097ms, 1h-pyrazole, 5-cyclopropyl
Formula Weight 108.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Cyclopropyl-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'