missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionyl Chloride, TRC

Product Code. 30359883
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359883 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359883 Supplier Toronto Research Chemicals Supplier No. TRCC98851050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Cyclopentylpropionyl Chloride
CAS 104-97-2
Molecular Formula C8H13ClO
Quantity 50 g
Synonym Cyclopentanepropionyl Chloride (8CI), 3-Cyclopentanepropionyl Chloride, 3-Cyclopentylpropanoyl Chloride, 3-Cyclopentylpropionic Acid Chloride, Cyclopentanepropanoyl Chloride
InChI Formula InChI=1S/C8H13ClO/c9-8(10)6-5-7-3-1-2-4-7/h7H,1-6H2
SMILES ClC(=O)CCC1CCCC1
IUPAC Name 3-cyclopentylpropanoyl chloride
Molecular Weight (g/mol) 160.64
Formula Weight 160.0655
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.