missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionic acid, 98%

Product Code. 11473717
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473717 100 g 100g
11463717 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473717 Supplier Thermo Scientific Alfa Aesar Supplier No. A18192.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140-77-2 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-77-2
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001392
InChI Key ZRPLANDPDWYOMZ-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
PubChem CID 8818
ChEBI CHEBI:50899
IUPAC Name 3-cyclopentylpropanoic acid
SMILES OC(=O)CCC1CCCC1

Specifications

Density 0.996
Boiling Point 130°C to 133°C (12 mmHg)
Flash Point 47°C (117°F)
Refractive Index 1.457
Quantity 100 g
UN Number UN1993
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material 3-Cyclopentylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.