missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyclopentyloxy)-4-methoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 10069391
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10069391 10 g Amber Glass Bottle 10g
10314442 1 g Amber Glass Bottle 1g
10490554 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10069391 Supplier Thermo Scientific Maybridge Supplier No. KM09079EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 144036-17-9
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.27
MDL Number MFCD00219797
InChI Key RVADCQWIQKYXBJ-UHFFFAOYSA-N
Synonym 3-cyclopentyloxy-4-methoxybenzoic acid, benzoic acid,3-cyclopentyloxy-4-methoxy, 3-cyclopentyloxy-4-methoxy benzoic acid, maybridge1_005004, acmc-20e1kk, 3-cyclopentyloxy-4-methoxy-benzoic acid, 3-cyclopentoxy-4-methoxy-benzoic acid, 3-cyclopentyloxy-4-methoxybezoic acid, 3-cyclopentyl-oxy-4-methoxybenzoic acid
PubChem CID 2779323
IUPAC Name 3-cyclopentyloxy-4-methoxybenzoic acid
SMILES COC1=C(OC2CCCC2)C=C(C=C1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 236.27
Percent Purity 97%
Chemical Name or Material 3-(Cyclopentyloxy)-4-methoxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.