missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexyl-1-propyne, 97%

Product Code. 11414517
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414517 1 g 1g
11424517 5 g 5g
11434517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414517 Supplier Thermo Scientific Alfa Aesar Supplier No. B20505.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17715-00-3 | C9H14 | 122.211 g/mol

3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17715-00-3
Molecular Formula C9H14
Molecular Weight (g/mol) 122.211
MDL Number MFCD00041562
InChI Key UARFKZSJGDQRLF-UHFFFAOYSA-N
Synonym 3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne
PubChem CID 87265
IUPAC Name prop-2-ynylcyclohexane
SMILES C#CCC1CCCCC1

Specifications

Density 0.845
Boiling Point 157°C to 158°C
Flash Point 35°C (95°F)
Refractive Index 1.462
Quantity 1 g
UN Number UN3295
Beilstein 1849402
Solubility Information Slightly soluble in water.
Formula Weight 122.21
Percent Purity 97%
Chemical Name or Material 3-Cyclohexyl-1-propyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.