missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexyl-1-propanol, 99%

Product Code. 10793241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10793241 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10793241 Supplier Thermo Scientific Acros Supplier No. 255880050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1124-63-6 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1124-63-6
CAS Min % 98.5
CAS Max % 100.0
Melting Point -60°C
Color Colorless
Density 0.9100g/mL
Boiling Point 223°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H18O
Linear Formula C6H11CH2CH2CH2OH
Refractive Index 1.4650 to 1.467
MDL Number MFCD00001528
Quantity 5 mL
Specific Gravity 0.91
Synonym cyclohexanepropanol, 3-cyclohexyl-1-propanol, 3-cyclohexylpropanol, unii-kjb3vo906f, kjb3vo906f, cyclohexylpropyl alcohol, 3-hydroxypropylcyclohexane, 3-cyclohexylpropyl alcohol, acmc-1c5ti
Solubility Information Solubility in water: 0.06% (20 c)
InChI Key CLYAQFSQLQTVNO-UHFFFAOYSA-N
SMILES C1CCC(CC1)CCCO
IUPAC Name 3-cyclohexylpropan-1-ol
Molecular Weight (g/mol) 142.24
PubChem CID 70763
Viscosity 36.7 mPa.s (20°C)
Formula Weight 142.24
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 3-Cyclohexyl-1-propanol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.