missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexenylcarboxylic Acid, TRC

Product Code. 30363729
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30363729

Brand: Toronto Research Chemicals TRCC98819510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Cyclohexenylcarboxylic Acid
CAS 4771-80-6
Molecular Formula C7H10O2
Quantity 10 g
Synonym (±)-3-Cyclohexene-1-carboxylic Acid, 1-Cyclohexene-4-carboxylic Acid, 3-Cyclohexenecarboxylic Acid, 3-Cyclohexenyl-1-formic Acid, 3-Cyclohexenylcarboxylic Acid, NSC 44712, NSC 44883, delta3-Cyclohexenecarboxylic Acid, delta3-Cyclohexenylcarboxylic Acid
InChI Formula InChI=1S/C7H10O2/c8-7(9)6-4-2-1-3-5-6/h1-2,6H,3-5H2,(H,8,9)
SMILES OC(=O)C1CCC=CC1
IUPAC Name cyclohex-3-ene-1-carboxylic acid
Molecular Weight (g/mol) 126.15
Formula Weight 126.0681
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.