missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyclohexenylcarboxylic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-Cyclohexenylcarboxylic Acid |
| CAS | 4771-80-6 |
| Molecular Formula | C7H10O2 |
| Quantity | 1 g |
| Synonym | (±)-3-Cyclohexene-1-carboxylic Acid, 1-Cyclohexene-4-carboxylic Acid, 3-Cyclohexenecarboxylic Acid, 3-Cyclohexenyl-1-formic Acid, 3-Cyclohexenylcarboxylic Acid, NSC 44712, NSC 44883, delta3-Cyclohexenecarboxylic Acid, delta3-Cyclohexenylcarboxylic Acid |
| InChI Formula | InChI=1S/C7H10O2/c8-7(9)6-4-2-1-3-5-6/h1-2,6H,3-5H2,(H,8,9) |
| SMILES | OC(=O)C1CCC=CC1 |
| IUPAC Name | cyclohex-3-ene-1-carboxylic acid |
| Molecular Weight (g/mol) | 126.15 |
| Formula Weight | 126.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction