missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexene-1-carbonyl chloride, 96%, Thermo Scientific™

Product Code. 11355379
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11355379

Brand: Thermo Scientific Alfa Aesar H50390.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Cyclohexene-1-carbonyl chloride is used as an active pharmaceutical ingredients.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-67-2
Molecular Formula C7H9ClO
Molecular Weight (g/mol) 144.60
MDL Number MFCD00013780
InChI Key CXWMHIRIXVNVQN-UHFFFAOYNA-N
Synonym 3-cyclohexene-1-carbonyl chloride, 3-cyclohexenecarbonyl chloride, cyclohex-3-enecarbonyl chloride, 3-cyclohexene-1-carbonylchloride, acmc-20an5a, 3-cyclohexenecarbonylchloride, 3-cyclohexene carbonyl chloride, 1-cyclohex-3-enecarbonyl chloride, 3-cyclohexene-1-carbonyl chloride #, 3-cyclohexenecarboxylic acid chloride
PubChem CID 523058
IUPAC Name cyclohex-3-ene-1-carbonyl chloride
SMILES ClC(=O)C1CCC=CC1

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 144.6
Percent Purity 96%
Chemical Name or Material 3-Cyclohexene-1-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.