missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine, 98%

Product Code. 10152230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10152230 100 g Glass bottle 100g
10540011 5 g Glass bottle 5g
10580351 500 g Glass bottle 500g
10485603 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10152230 Supplier Thermo Scientific Acros Supplier No. 110861000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-54-9 | C6H4N2 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-54-9
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00006372
InChI Key GZPHSAQLYPIAIN-UHFFFAOYSA-N
Synonym 3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
PubChem CID 79
ChEBI CHEBI:86556
IUPAC Name pyridine-3-carbonitrile
SMILES N#CC1=CC=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 47.0°C to 51.0°C
Color Colorless or White to Yellow
Density 1.1590g/mL
Boiling Point 201.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CN
Quantity 100 g
Beilstein 22, 41
Specific Gravity 1.159
Solubility Information Solubility in water: 140g/L (20°C)
Formula Weight 104.11
Percent Purity 98%
Physical Form Crystalline Mass, Crystals or Chunks
Chemical Name or Material 3-Cyanopyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Cyanopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.