missing translation for 'onlineSavingsMsg'
Learn More

(3-Cyanopropyl)triethoxysilane, 97%

Product Code. 11317425
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317425 25 g 25g
11327425 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317425 Supplier Thermo Scientific Alfa Aesar Supplier No. L05367.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1067-47-6 | C10H21NO3Si | 231.367 g/mol

(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.

Solubility
Not miscible in water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-47-6
Molecular Formula C10H21NO3Si
Molecular Weight (g/mol) 231.367
MDL Number MFCD00042636
InChI Key VGIURMCNTDVGJM-UHFFFAOYSA-N
Synonym 3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech
PubChem CID 14024
IUPAC Name 4-triethoxysilylbutanenitrile
SMILES CCO[Si](CCCC#N)(OCC)OCC

Specifications

Boiling Point 238°C to 241°C
Quantity 25 g
Linear Formula NCCH2CH2CH2Si(OCH2CH3)3
Beilstein 1942237
Flash Point 74°C (165°F)
Solubility Information Not miscible in water.
Formula Weight 231.37
Percent Purity 97%
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.961
Chemical Name or Material (3-Cyanopropyl)triethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • (3-Cyanopropyl)triethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: (3-Cyanopropyl)triethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.