missing translation for 'onlineSavingsMsg'
Learn More

(3-Cyanopropyl)dimethylchlorosilane, 94%

Product Code. 15448819
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448819 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448819 Supplier Thermo Scientific Alfa Aesar Supplier No. L03607.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18156-15-5 | C6H12ClNSi | 161.704 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 108°C to 109°C (15mmHg)
CAS 18156-15-5
Quantity 10 g
Molecular Formula C6H12ClNSi
MDL Number MFCD00001969
UN Number UN2987
Beilstein 2235062
Flash Point 85°C (185°F)
Synonym 3-cyanopropyl dimethylchlorosilane, 4-chlorodimethylsilyl butanenitrile, 3-cyanopropyldimethylchlorosilane, chloro 3-cyanopropyl dimethylsilane, butanenitrile, 4-chlorodimethylsilyl, 4-chlorodimethylsilyl butyronitrile, 4-chloro dimethyl silyl butanenitrile, cyanopropyldimethylchlorosilane, chlorocyanopropyldimethylsilane, cyanopropyl dimethylchlorosilane
InChI Key GPIARXZSVWTOMD-UHFFFAOYSA-N
SMILES C[Si](C)(CCCC#N)Cl
IUPAC Name 4-[chloro(dimethyl)silyl]butanenitrile
Molecular Weight (g/mol) 161.704
PubChem CID 87478
Formula Weight 161.71
Percent Purity 94%
Refractive Index 1.446
Sensitivity Moisture sensitive
Density 0.99
Chemical Name or Material (3-Cyanopropyl)dimethylchlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.