missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenyl isothiocyanate, 97%

Product Code. 11357856
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357856 1 g 1g
11367856 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357856 Supplier Thermo Scientific Alfa Aesar Supplier No. L13045.03
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3125-78-8 | C8H4N2S | 160.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3125-78-8
Molecular Formula C8H4N2S
Molecular Weight (g/mol) 160.19
MDL Number MFCD00041070
InChI Key UXCYPHCFQAFVFH-UHFFFAOYSA-N
Synonym 3-cyanophenyl isothiocyanate, benzonitrile, 3-isothiocyanato, m-cyanophenyl isothiocyanate, 3-cyanophenylisothiocyanate, acmc-1bn6o, benzonitrile,3-isothiocyanato, 3-isothiocyanatobenzenecarbonitrile
PubChem CID 76578
IUPAC Name 3-isothiocyanatobenzonitrile
SMILES S=C=NC1=CC=CC(=C1)C#N

Specifications

Melting Point 64°C to 66°C
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2803099
Sensitivity Moisture sensitive
Formula Weight 160.2
Percent Purity 97%
Chemical Name or Material 3-Cyanophenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.